missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxyphenol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 450-93-1
Molecular Formula C7H7FO2
Molecular Weight (g/mol) 142.13
MDL Number MFCD00070797
InChI Key OULGLTLTWBZBLO-UHFFFAOYSA-N
Synonym 4-fluoroguaiacol, 5-fluoro-2-hydroxyanisole, 4-fluoro-2-methoxy-phenol, phenol, 4-fluoro-2-methoxy, 4-fluoro-2-methyoxyphenol, pubchem3880, acmc-1agod, 2-methoxy-4-fluorophenol, 4-fluoro-2-methoxyphenol
PubChem CID 2737368
IUPAC Name 4-fluoro-2-methoxyphenol
SMILES COC1=CC(F)=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398990010
SDS View Documents
Thermo Scientific Chemicals
398990010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398990050
SDS View Documents
Thermo Scientific Chemicals
398990050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

450-93-1
142.13
OULGLTLTWBZBLO-UHFFFAOYSA-N
2737368
COC1=CC(F)=CC=C1O
C7H7FO2
MFCD00070797
4-fluoroguaiacol, 5-fluoro-2-hydroxyanisole, 4-fluoro-2-methoxy-phenol, phenol, 4-fluoro-2-methoxy, 4-fluoro-2-methyoxyphenol, pubchem3880, acmc-1agod, 2-methoxy-4-fluorophenol, 4-fluoro-2-methoxyphenol
4-fluoro-2-methoxyphenol

Specifications

450-93-1
100.0
195.0°C
Authentic
Glass bottle
1.5160 to 1.5180 (20°C, 589nm)
MFCD00070797
1.247
OULGLTLTWBZBLO-UHFFFAOYSA-N
4-fluoro-2-methoxyphenol
2737368
97%
96.0
1.2470g/mL
101°C
96% min. (GC)
C7H7FO2
FC6H3(OH)OCH3
1g
4-fluoroguaiacol, 5-fluoro-2-hydroxyanisole, 4-fluoro-2-methoxy-phenol, phenol, 4-fluoro-2-methoxy, 4-fluoro-2-methyoxyphenol, pubchem3880, acmc-1agod, 2-methoxy-4-fluorophenol, 4-fluoro-2-methoxyphenol
COC1=CC(F)=CC=C1O
142.13
142.13
4-Fluoro-2-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only