missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-methoxyacetophenone, 97%

Catalog No. AAL0986203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0986203 1 g
AAL0986206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0986203 Supplier Thermo Scientific Chemicals Supplier No. L0986203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51788-80-8 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51788-80-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00061143
InChI Key YOXBPWVWNQROBJ-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-methoxyphenyl ethanone, 4-fluoro-2-methoxyacetophenone, 4'-fluoro-2'-methoxyacetophenone, 1-4-fluoro-2-methoxyphenyl ethan-1-one, 2'-methoxy-4'-fluoroacetophenone, 1-4-fluoro-2-methoxy-phenyl-ethanone, 1-acetyl-4-fluoro-2-methoxybenzene, 2-methoxy-4-fluoroacetophenone, 4'-fluoro-2'-methoxyacetophenone,, pubchem13459
PubChem CID 142887
IUPAC Name 1-(4-fluoro-2-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)OC

Specifications

Melting Point 51°C to 53°C
Quantity 1 g
Beilstein 2251986
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-2'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only