missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-hydroxyacetophenone, 98%

Catalog No. AAA1603006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1603006 5 g
AAA1603014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1603006 Supplier Thermo Scientific Chemicals Supplier No. A1603006
Only null left

CAS: 1481-27-2 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1481-27-2
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143203
InChI Key HLTBTUXAMVOKIH-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl, 1-4-fluoro-2-hydroxy-phenyl ethanone, 1-acetyl-4-fluoro-2-hydroxybenzene, pubchem13252, acmc-1bzss, 4'-fluoro-2'-hydroxy
PubChem CID 2737326
IUPAC Name 1-(4-fluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C=C1O

Specifications

Melting Point 29°C to 34°C
Boiling Point 65°C (2 mmHg)
Flash Point 98°C (208°F)
Quantity 5 g
Beilstein 2249799
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4'-Fluoro-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only