missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,1,3-benzoxadiazole 98.0+%, TCI America™

Catalog No. p-7129841
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07021G 1 g
F07025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07021G Supplier TCI America Supplier No. F07021G
Only null left

Chemical Identifiers

CAS 29270-55-1
Molecular Formula C6H3FN2O
Molecular Weight (g/mol) 138.101
MDL Number MFCD03425566
InChI Key QKZQJBBZKDVVFI-UHFFFAOYSA-N
Synonym 4-Fluorobenzofurazan
PubChem CID 4607136
IUPAC Name 4-fluoro-2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C(=C1)F

Specifications

Boiling Point 83°C
Quantity 1 g
Formula Weight 138.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2,1,3-benzoxadiazole