missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7130056
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1080200MG 200 mg
F10801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1080200MG Supplier TCI America Supplier No. F1080200MG
Only null left

Chemical Identifiers

CAS 182344-25-8
Molecular Formula C10H8BFO2
Molecular Weight (g/mol) 189.98
MDL Number MFCD04039221
InChI Key IDCGAEJGZLYEAZ-UHFFFAOYSA-N
Synonym 4-fluoronaphthalene-1-boronic acid, 4-fluoronaphtalene-1-boronic acid, 4-fluoronaphthalen-1-yl-1-boronic acid, 4-fluoronaphthalen-1-yl boronic acid, 4-fluoro-1-naphthyl boronic acid, 4-fluoro-1-naphthaleneboronic acid, boronic acid, 4-fluoro-1-naphthalenyl, akos brn-0210, acmc-209ejb, 4-fluoronaphth-1-ylboronic acid
PubChem CID 2782675
IUPAC Name (4-fluoronaphthalen-1-yl)boronic acid
SMILES B(C1=CC=C(C2=CC=CC=C12)F)(O)O

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 200 mg
Formula Weight 189.98
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-1-naphthaleneboronic Acid (contains varying amounts of Anhydride)