missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-iodo-2-nitrobenzene 98.0+%, TCI America™

Catalog No. F08285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08285G Supplier TCI America Supplier No. F08285G
Only null left

Specifications

CAS 364-77-2
Boiling Point 128°C
Molecular Formula C6H3FINO2
Quantity 5 g
MDL Number MFCD00153169
Synonym 2-iodo-5-fluoronitrobenzene, 5-fluoro-2-iodonitrobenzene, 4-fluoro-2-nitroiodobenzene, 1-fluoro-4-iodo-3-nitrobenzene, benzene, 4-fluoro-1-iodo-2-nitro, pubchem1058, acmc-209y4c, ksc495i0t, 1-iodo-2-nitro-4-fluorobenzene, buttpark 29\07-56
InChI Key ANPFMDLUEWNKIM-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC(F)=CC=C1I
IUPAC Name 4-fluoro-1-iodo-2-nitrobenzene
Molecular Weight (g/mol) 267.00
PubChem CID 2737347
Formula Weight 267.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-1-iodo-2-nitrobenzene
Show More Show Less
TSCA No
Recommended Storage Refrigerator