missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,3-dioxolan-2-one 98.0+%, TCI America™

Catalog No. p-7129862
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07315G 5 g
F073125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07315G Supplier TCI America Supplier No. F07315G

Chemical Identifiers

CAS 114435-02-8
Molecular Formula C3H3FO3
Molecular Weight (g/mol) 106.052
MDL Number MFCD06247543
InChI Key SBLRHMKNNHXPHG-UHFFFAOYSA-N
Synonym fluoroethylene carbonate, 1,3-dioxolan-2-one, 4-fluoro, fluoro ethylene carbonate, 4-fluoro-1,3-dioxalan-2-one, 4-fluoro-2-oxo-1,3-dioxolane, 4-fluoro-ethylene carbonate, 4-fluoro-ethylene carboante, ksc219e1h, fec, lithium ion battery grade
PubChem CID 2769656
IUPAC Name 4-fluoro-1,3-dioxolan-2-one
SMILES C1C(OC(=O)O1)F

Specifications

Melting Point 20°C
Color Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 106.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-1,3-dioxolan-2-one
EINECSNumber (5)-6911&(5)-6953
TSCA Yes
Recommended Storage Refrigerator