missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-1,3-dioxolan-2-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 20°C |
| Color | Yellow |
| Boiling Point | 210°C |
| Quantity | 25 g |
| Formula Weight | 106.05 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Fluoro-1,3-dioxolan-2-one |
Safety and Handling
| EINECSNumber | (5)-6911&(5)-6953 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |