missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynyltoluene 98.0+%, TCI America™

Catalog No. p-7129088
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06555G 5 g
E065525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06555G Supplier TCI America Supplier No. E06555G

Chemical Identifiers

CAS 766-97-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
InChI Key KSZVOXHGCKKOLL-UHFFFAOYSA-N
Synonym 4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
PubChem CID 13018
IUPAC Name 1-ethynyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C#C

Specifications

Boiling Point 168°C
Quantity 5 g
UN Number 1325
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Ethynyltoluene
RTECSNumber XT2570000
TSCA No