missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethynylpyridine Hydrochloride 95.0+%, TCI America™
$123.21 - $365.79
Chemical Identifiers
| CAS | 352530-29-1 |
|---|---|
| Molecular Formula | C7H6ClN |
| Molecular Weight (g/mol) | 139.582 |
| MDL Number | MFCD03093912 |
| InChI Key | KFBZWZGIZHLUBX-UHFFFAOYSA-N |
| Synonym | 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1 |
| PubChem CID | 16216931 |
| IUPAC Name | 4-ethynylpyridine;hydrochloride |
| SMILES | C#CC1=CC=NC=C1.Cl |
Chemical Identifiers
| 352530-29-1 | |
| 139.582 | |
| KFBZWZGIZHLUBX-UHFFFAOYSA-N | |
| 16216931 | |
| C#CC1=CC=NC=C1.Cl |
| C7H6ClN | |
| MFCD03093912 | |
| 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1 | |
| 4-ethynylpyridine;hydrochloride |
Specifications
| 352530-29-1 | |
| Green-Yellow | |
| MFCD03093912 | |
| 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1 | |
| C#CC1=CC=NC=C1.Cl | |
| 139.582 | |
| 139.58 | |
| Crystalline Powder |
| 150°C | |
| C7H6ClN | |
| 1 g | |
| KFBZWZGIZHLUBX-UHFFFAOYSA-N | |
| 4-ethynylpyridine;hydrochloride | |
| 16216931 | |
| ≥95.0% (HPLC,T) | |
| 4-Ethynylpyridine Hydrochloride |
Safety and Handling
TSCA : No