missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethynylpyridine Hydrochloride 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 150°C |
| Color | Green-Yellow |
| Quantity | 5 g |
| Formula Weight | 139.58 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Ethynylpyridine Hydrochloride |
Safety and Handling
| TSCA | No |