missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylpyridine Hydrochloride 95.0+%, TCI America™

Catalog No. E10431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10431G 1 g
E10435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10431G Supplier TCI America Supplier No. E10431G

Chemical Identifiers

CAS 352530-29-1
Molecular Formula C7H6ClN
Molecular Weight (g/mol) 139.582
MDL Number MFCD03093912
InChI Key KFBZWZGIZHLUBX-UHFFFAOYSA-N
Synonym 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1
PubChem CID 16216931
IUPAC Name 4-ethynylpyridine;hydrochloride
SMILES C#CC1=CC=NC=C1.Cl

Specifications

Melting Point 150°C
Color Green-Yellow
Quantity 1 g
Formula Weight 139.58
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylpyridine Hydrochloride