missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylpyridine hydrochloride, 97%

Catalog No. AC428490010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428490010 1 g Glass bottle
AC428490050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428490010 Supplier Thermo Scientific Chemicals Supplier No. 428490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 352530-29-1 | C7H5N·ClH | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 352530-29-1
Molecular Formula C7H5N·ClH
Molecular Weight (g/mol) 139.58
MDL Number MFCD03093912
InChI Key KFBZWZGIZHLUBX-UHFFFAOYSA-N
Synonym 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1
PubChem CID 16216931
IUPAC Name 4-ethynylpyridine;hydrochloride
SMILES C#CC1=CC=NC=C1.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 139.58
Percent Purity 97%
Chemical Name or Material 4-Ethynylpyridine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only