missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylphthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7129074
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05791G 1 g
E05795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05791G Supplier TCI America Supplier No. E05791G

Chemical Identifiers

CAS 73819-76-8
Molecular Formula C10H4O3
Molecular Weight (g/mol) 172.14
MDL Number MFCD06200801
InChI Key DXJLXGJIZZNCBO-UHFFFAOYSA-N
PubChem CID 11321228
IUPAC Name 5-ethynyl-1,3-dihydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1C=CC(=C2)C#C

Specifications

Melting Point 125°C
Quantity 1 g
Formula Weight 172.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylphthalic Anhydride