missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylaniline 98.0+%, TCI America™

Catalog No. p-7129034
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E050510G 10 g
E050525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E050510G Supplier TCI America Supplier No. E050510G
Only null left

Chemical Identifiers

CAS 14235-81-5
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00168824
InChI Key JXYITCJMBRETQX-UHFFFAOYSA-N
Synonym 4-aminophenylacetylene, 4-ethynyl aniline, benzenamine, 4-ethynyl, p-aminophenylacetylene, 1-amino-4-ethynylbenzene, 4-aminophenyl acetylene, p-ethynylaniline, 4-ethynylaniline, 4-ethynyl-phenylamine, p-aminophenyl-acetylene
PubChem CID 3760025
IUPAC Name 4-ethynylaniline
SMILES C#CC1=CC=C(C=C1)N

Specifications

Melting Point 104°C
Quantity 10 g
Formula Weight 117.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylaniline