missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynyl-4'-propylbiphenyl 98.0+%, TCI America™

Catalog No. E12311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12311G 1 g
E12315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12311G Supplier TCI America Supplier No. E12311G
Only null left

Chemical Identifiers

CAS 360768-57-6
Molecular Formula C17H16
Molecular Weight (g/mol) 220.315
MDL Number MFCD18072535
InChI Key ZBSZCHDBZRCUES-UHFFFAOYSA-N
PubChem CID 19814911
IUPAC Name 1-ethynyl-4-(4-propylphenyl)benzene
SMILES CCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 220.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynyl-4′-propylbiphenyl