missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyltoluene, 97%

Catalog No. AAL0212006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0212006 5 g
AAL0212014 25 g
AAL0212022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0212006 Supplier Thermo Scientific Chemicals Supplier No. L0212006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 622-96-8 | C9H12 | 120.195 g/mol

4-Ethyltoluene is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethyltoluene is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 622-96-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009263
InChI Key JRLPEMVDPFPYPJ-UHFFFAOYSA-N
Synonym 4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
PubChem CID 12160
IUPAC Name 1-ethyl-4-methylbenzene
SMILES CCC1=CC=C(C=C1)C

Specifications

Melting Point -62°C
Density 0.861
Boiling Point 161°C to 163°C
Flash Point 43°C (109°F)
Refractive Index 1.495
Quantity 5 g
UN Number UN3295
Beilstein 1903007
Solubility Information Insoluble in water.
Formula Weight 120.2
Percent Purity 97%
Chemical Name or Material 4-Ethyltoluene
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 210-761-2
RTECSNumber XT2550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only