missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$54.06 - $54.06
Chemical Identifiers
| CAS | 63139-21-9 |
|---|---|
| Molecular Formula | C8H11BO2 |
| Molecular Weight (g/mol) | 149.98 |
| MDL Number | MFCD00859377 |
| InChI Key | RZCPLOMUUCFPQA-UHFFFAOYSA-N |
| PubChem CID | 2734352 |
| IUPAC Name | (4-ethylphenyl)boronic acid |
| SMILES | CCC1=CC=C(C=C1)B(O)O |
Chemical Identifiers
| 63139-21-9 | |
| 149.98 | |
| RZCPLOMUUCFPQA-UHFFFAOYSA-N | |
| (4-ethylphenyl)boronic acid |
| C8H11BO2 | |
| MFCD00859377 | |
| 2734352 | |
| CCC1=CC=C(C=C1)B(O)O |
Specifications
| 63139-21-9 | |
| C8H11BO2 | |
| 5 g | |
| RZCPLOMUUCFPQA-UHFFFAOYSA-N | |
| (4-ethylphenyl)boronic acid | |
| 2734352 | |
| Crystalline Powder |
| 141°C | |
| MFCD00859377 | |
| 4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6 | |
| CCC1=CC=C(C=C1)B(O)O | |
| 149.98 | |
| 149.98 | |
| 4-Ethylphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No