missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E07205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07205G Supplier TCI America Supplier No. E07205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63139-21-9
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD00859377
InChI Key RZCPLOMUUCFPQA-UHFFFAOYSA-N
PubChem CID 2734352
IUPAC Name (4-ethylphenyl)boronic acid
SMILES CCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 141°C
Quantity 5 g
Synonym 4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
Formula Weight 149.98
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethylphenylboronic Acid (contains varying amounts of Anhydride)