missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylacetylene, 99%, Thermo Scientific Chemicals

Catalog No. AAH2631306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2631306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2631306 Supplier Thermo Scientific Chemicals Supplier No. H2631306

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40307-11-7
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00173887
InChI Key ZNTJVJSUNSUMPP-UHFFFAOYSA-N
Synonym 4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
PubChem CID 142425
IUPAC Name 1-ethyl-4-ethynylbenzene
SMILES CCC1=CC=C(C=C1)C#C

Specifications

Density 0.93
Boiling Point 192°C
Flash Point 71°C (160°F)
Refractive Index 1.542
Quantity 5 g
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material 4-Ethylphenylacetylene
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only