missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl Isocyanate 97.0+%, TCI America™

$50.05 - $134.75

Chemical Identifiers

CAS 23138-50-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013881
InChI Key FWPYUSLQCQDLJR-UHFFFAOYSA-N
Synonym 4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
PubChem CID 90007
IUPAC Name 1-ethyl-4-isocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E07671G
View Documents
TCI America
E07671G
1 g N/A
Only null left
 
E07675G
View Documents
TCI America
E07675G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

23138-50-3
147.177
FWPYUSLQCQDLJR-UHFFFAOYSA-N
90007
CCC1=CC=C(C=C1)N=C=O
C9H9NO
MFCD00013881
4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
1-ethyl-4-isocyanatobenzene

Specifications

23138-50-3
80°C
MFCD00013881
2206
FWPYUSLQCQDLJR-UHFFFAOYSA-N
1-ethyl-4-isocyanatobenzene
90007
≥97.0% (GC)
4-Ethylphenyl Isocyanate
Yellow
C9H9NO
1 g
4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
CCC1=CC=C(C=C1)N=C=O
147.177
147.18
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'