missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl Isocyanate 97.0+%, TCI America™

Catalog No. E07671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07671G 1 g
E07675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07671G Supplier TCI America Supplier No. E07671G

Chemical Identifiers

CAS 23138-50-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013881
InChI Key FWPYUSLQCQDLJR-UHFFFAOYSA-N
Synonym 4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
PubChem CID 90007
IUPAC Name 1-ethyl-4-isocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
UN Number 2206
Formula Weight 147.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylphenyl Isocyanate