Learn More
4-Ethylphenyl isocyanate, 98%
CAS: 23138-50-3 | C9H9NO | 147.177 g/mol
Supplier: Thermo Scientific Chemicals L1271806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Ethylphenyl isocyanate | |
| 1.024 | |
| 78°C (172°F) | |
| C9H9NO | |
| MFCD00013881 | |
| UN2206 | |
| Moisture sensitive | |
| FWPYUSLQCQDLJR-UHFFFAOYSA-N | |
| 1-ethyl-4-isocyanatobenzene | |
| 90007 | |
| 98% |
| 23138-50-3 | |
| 67°C to 68°C (1 mmHg) | |
| Pungent | |
| 1.527 | |
| 5 g | |
| 971100 | |
| 4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene | |
| CCC1=CC=C(C=C1)N=C=O | |
| 147.177 | |
| 147.18 |
Chemical Identifiers
| 23138-50-3 | |
| 147.177 | |
| FWPYUSLQCQDLJR-UHFFFAOYSA-N | |
| 1-ethyl-4-isocyanatobenzene |
| C9H9NO | |
| MFCD00013881 | |
| 90007 | |
| CCC1=CC=C(C=C1)N=C=O |
Safety and Handling
GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H301-H315-H317-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 245-446-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only