missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl isocyanate, 98%

Catalog No. AAL1271814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1271814 25 g
AAL1271803 1 g
AAL1271806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1271814 Supplier Thermo Scientific Chemicals Supplier No. L1271814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23138-50-3 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23138-50-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013881
InChI Key FWPYUSLQCQDLJR-UHFFFAOYSA-N
PubChem CID 90007
IUPAC Name 1-ethyl-4-isocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=O

Specifications

Density 1.024
Boiling Point 67°C to 68°C (1 mmHg)
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.527
Quantity 25 g
UN Number UN2206
Beilstein 971100
Sensitivity Moisture sensitive
Synonym 4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 4-Ethylphenyl isocyanate
Show More Show Less
Hazard Category H227-H301-H315-H317-H319-H330-H335
Hazard Statement GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 245-446-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only