missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylmorpholine 99.0+%, TCI America™

Catalog No. p-7128862
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E014525ML 25 mL
E0145500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E014525ML Supplier TCI America Supplier No. E014525ML
Only null left

Chemical Identifiers

CAS 100-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00006177
InChI Key HVCNXQOWACZAFN-UHFFFAOYSA-N
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
PubChem CID 7525
IUPAC Name 4-ethylmorpholine
SMILES CCN1CCOCC1

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 139°C
Quantity 25 mL
UN Number 2734
Formula Weight 115.18
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Ethylmorpholine
EINECSNumber (5)-0860
RTECSNumber QE4025000
TSCA Yes