missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylmorpholine, 98%

Catalog No. AAA11905AU
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11905AU 1000 mL
AAA11905AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11905AU Supplier Thermo Scientific Chemicals Supplier No. A11905AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-74-3 | C6H13NO | 115.176 g/mol

N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

Solubility
Miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 100-74-3
Molecular Formula C6H13NO
MDL Number MFCD00006177
InChI Key HVCNXQOWACZAFN-UHFFFAOYSA-N
PubChem CID 7525
IUPAC Name 4-ethylmorpholine
SMILES CCN1CCOCC1

Specifications

Melting Point -63°C
Density 0.908
Boiling Point 138°C to 139°C
Flash Point 27°C (80°F)
Refractive Index 1.4413
Quantity 1000 mL
UN Number UN2734
Beilstein 102969
Sensitivity Air Sensitive
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
Solubility Information Miscible with water.
Molecular Weight (g/mol) 115.176
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 4-Ethylmorpholine
Show More Show Less
Hazard Category H226-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 202-885-0
RTECSNumber QE4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only