missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcyclohexanone, 99%

Catalog No. AAL0936314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0936314 25 g
AAL0936322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0936314 Supplier Thermo Scientific Chemicals Supplier No. L0936314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5441-51-0 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5441-51-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00001644
InChI Key OKSDJGWHKXFVME-UHFFFAOYSA-N
Synonym 4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl
PubChem CID 79506
IUPAC Name 4-ethylcyclohexan-1-one
SMILES CCC1CCC(=O)CC1

Specifications

Density 0.895
Boiling Point 192°C to 194°C
Flash Point 63°C (145°F)
Odor Characteristic
Refractive Index 1.4515
Quantity 25 g
Beilstein 1924964
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 4-Ethylcyclohexanone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 226-629-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only