missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzeneboronic acid, 97%

CAS: 63139-21-9 | C8H11BO2 | 149.98 g/mol

$48.55 - $504.88

Chemical Identifiers

CAS 63139-21-9
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD00859377
InChI Key RZCPLOMUUCFPQA-UHFFFAOYSA-N
Synonym 4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
PubChem CID 2734352
IUPAC Name (4-ethylphenyl)boronic acid
SMILES CCC1=CC=C(C=C1)B(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2465603
View Documents
Thermo Scientific Chemicals
B2465603
1 g
Each for $48.55
Only null left
 
AAB2465606
View Documents
Thermo Scientific Chemicals
B2465606
5 g
Each for $142.27
Only null left
 
AAB2465614
View Documents
Thermo Scientific Chemicals
B2465614
25 g
Each for $504.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63139-21-9
149.98
RZCPLOMUUCFPQA-UHFFFAOYSA-N
2734352
CCC1=CC=C(C=C1)B(O)O
C8H11BO2
MFCD00859377
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
(4-ethylphenyl)boronic acid

Specifications

63139-21-9
C8H11BO2
1 g
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
CCC1=CC=C(C=C1)B(O)O
149.98
149.99
4-Ethylbenzeneboronic acid
150°C to 154°C
MFCD00859377
3030638
RZCPLOMUUCFPQA-UHFFFAOYSA-N
(4-ethylphenyl)boronic acid
2734352
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only