missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylamino-1-butanol 98.0+%, TCI America™

Catalog No. p-7129018
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04735G 5 g
E047325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04735G Supplier TCI America Supplier No. E04735G

Chemical Identifiers

CAS 39216-86-9
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00671534
InChI Key PVNNOLUAMRODAC-UHFFFAOYSA-N
Synonym N-Ethylbutanolamine
PubChem CID 546882
IUPAC Name 4-(ethylamino)butan-1-ol
SMILES CCNCCCCO

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 117.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Ethylamino-1-butanol