missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethyl-4'-(trans-4-pentylcyclohexyl)biphenyl 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 334.55 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Ethyl-4′-(trans-4-pentylcyclohexyl)biphenyl |
Safety and Handling
| TSCA | No |