missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-4'-(trans-4-pentylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. E12541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12541G 1 g
E12545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12541G Supplier TCI America Supplier No. E12541G
Only null left

Chemical Identifiers

CAS 79709-85-6
Molecular Formula C25H34
Molecular Weight (g/mol) 334.55
MDL Number MFCD01941072
InChI Key JOLGXBQYTARJLD-UHFFFAOYSA-N
Synonym trans-4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-pentylcyclohexyl phenyl benzene, 4-ethyl-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 1-4-ethylphenyl-4-4-pentylcyclohexyl benzene, 1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl, bch-52, bpch-52, 4-ethyl-4'-4-pentylcyclohexyl biphenyl, 4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-n-pentylcyclohexyl biphenyl
PubChem CID 630553
IUPAC Name 4-ethyl-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
SMILES CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 334.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethyl-4′-(trans-4-pentylcyclohexyl)biphenyl