missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl Isocyanate 98.0+%, TCI America™

Catalog No. E07705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07705G Supplier TCI America Supplier No. E07705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32459-62-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00013877
InChI Key FMYVTFRADSNGDN-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
PubChem CID 122887
IUPAC Name 1-ethoxy-4-isocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=O

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2206
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethoxyphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator