missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isocyanate, 97%

CAS: 32459-62-4 | C9H9NO2 | 163.18 g/mol

$79.79 - $234.78

Chemical Identifiers

CAS 32459-62-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00013877
InChI Key FMYVTFRADSNGDN-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
PubChem CID 122887
IUPAC Name 1-ethoxy-4-isocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1150906
View Documents
Thermo Scientific Chemicals
L1150906
5 g
Each for $79.79
Only null left
 
AAL1150914
View Documents
Thermo Scientific Chemicals
L1150914
25 g
Each for $234.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32459-62-4
163.18
FMYVTFRADSNGDN-UHFFFAOYSA-N
122887
CCOC1=CC=C(C=C1)N=C=O
C9H9NO2
MFCD00013877
4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
1-ethoxy-4-isocyanatobenzene

Specifications

32459-62-4
235°C
C9H9NO2
MFCD00013877
UN2206
Moisture sensitive; May form a precipitate
FMYVTFRADSNGDN-UHFFFAOYSA-N
1-ethoxy-4-isocyanatobenzene
122887
97%
1.11
105°C (221°F)
1.536
5 g
742647
4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
CCOC1=CC=C(C=C1)N=C=O
163.18
163.18
4-Ethoxyphenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c

H302+H332-H315-H319-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only