missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl 4-Butylbenzoate 99.0+%, TCI America™

Catalog No. B03751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B03751G Supplier TCI America Supplier No. B03751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62716-65-8
Molecular Formula C19H22O3
Molecular Weight (g/mol) 298.382
MDL Number MFCD00059426
InChI Key DDDWTNWRNNRWNQ-UHFFFAOYSA-N
Synonym 4-Butylbenzoic Acid 4-Ethoxyphenyl Ester
PubChem CID 595107
IUPAC Name (4-ethoxyphenyl) 4-butylbenzoate
SMILES CCCCC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)OCC

Specifications

Melting Point 63°C
Color White
Boiling Point 237°C
Quantity 1 g
Formula Weight 298.38
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxyphenyl 4-Butylbenzoate