missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxymethyl)-2-methoxyphenol 98.0+%, TCI America™

Catalog No. E10285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10285G Supplier TCI America Supplier No. E10285G
Only null left

Chemical Identifiers

CAS 13184-86-6
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD06409999
InChI Key KOCVACNWDMSLBM-UHFFFAOYSA-N
Synonym Ethyl Vanillyl Ether
PubChem CID 61586
IUPAC Name 4-(ethoxymethyl)-2-methoxyphenol
SMILES CCOCC1=CC(=C(C=C1)O)OC

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Ethoxymethyl)-2-methoxyphenol