missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycinnamic Acid 98.0+%, TCI America™

Catalog No. p-7129186
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08565G 5 g
E085625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08565G Supplier TCI America Supplier No. E08565G
Only null left

Chemical Identifiers

CAS 2373-79-7
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD11040581
InChI Key DZLOUWYGNATKKZ-VMPITWQZSA-N
Synonym 4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239
PubChem CID 704218
IUPAC Name (E)-3-(4-ethoxyphenyl)prop-2-enoic acid
SMILES CCOC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 199°C
Color Yellow
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxycinnamic Acid