missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycinnamic acid, prediminantly trans, 98+%

Catalog No. p-7044417
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0632406 5 g
AAL0632414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0632406 Supplier Thermo Scientific Chemicals Supplier No. L0632406
Only null left

CAS: 2373-79-7 | C11H12O3 | 192.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2373-79-7
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00016848
InChI Key DZLOUWYGNATKKZ-VMPITWQZSA-N
Synonym 4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239
PubChem CID 704218
IUPAC Name (E)-3-(4-ethoxyphenyl)prop-2-enoic acid
SMILES CCOC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 195°C to 199°C
Quantity 5 g
Beilstein 3198068
Formula Weight 192.22
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only