missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$65.46 - $65.46
Chemical Identifiers
| CAS | 4334-88-7 |
|---|---|
| Molecular Formula | C9H11BO4 |
| Molecular Weight (g/mol) | 193.99 |
| MDL Number | MFCD02179441 |
| InChI Key | ZLNFACCFYUFTLD-UHFFFAOYSA-N |
| PubChem CID | 2734350 |
| IUPAC Name | [4-(ethoxycarbonyl)phenyl]boronic acid |
| SMILES | CCOC(=O)C1=CC=C(C=C1)B(O)O |
Chemical Identifiers
| 4334-88-7 | |
| 193.99 | |
| ZLNFACCFYUFTLD-UHFFFAOYSA-N | |
| [4-(ethoxycarbonyl)phenyl]boronic acid |
| C9H11BO4 | |
| MFCD02179441 | |
| 2734350 | |
| CCOC(=O)C1=CC=C(C=C1)B(O)O |
Specifications
| 4334-88-7 | |
| White | |
| MFCD02179441 | |
| 4-ethoxycarbonylphenyl boronic acid, 4-ethoxycarbonyl phenylboronic acid, 4-ethoxycarbonyl phenyl boronic acid, 4-ethoxycarbonyl benzeneboronic acid, 4-ethoxycarbonylbenzeneboronic acid, ethyl 4-boronobenzoate, p-ethoxycarbonylphenylboronic acid, ethyl 4-dihydroxyboranyl benzoate, 4-carbethoxyphenylboronic acid, benzoic acid, 4-borono-, 1-ethyl ester | |
| CCOC(=O)C1=CC=C(C=C1)B(O)O | |
| 193.99 | |
| 193.99 | |
| 4-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
| 160°C | |
| C9H11BO4 | |
| 1 g | |
| ZLNFACCFYUFTLD-UHFFFAOYSA-N | |
| [4-(ethoxycarbonyl)phenyl]boronic acid | |
| 2734350 | |
| Crystalline Powder |
Safety and Handling
TSCA : No