missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E08681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08681G Supplier TCI America Supplier No. E08681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4334-88-7
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179441
InChI Key ZLNFACCFYUFTLD-UHFFFAOYSA-N
PubChem CID 2734350
IUPAC Name [4-(ethoxycarbonyl)phenyl]boronic acid
SMILES CCOC(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Synonym 4-ethoxycarbonylphenyl boronic acid, 4-ethoxycarbonyl phenylboronic acid, 4-ethoxycarbonyl phenyl boronic acid, 4-ethoxycarbonyl benzeneboronic acid, 4-ethoxycarbonylbenzeneboronic acid, ethyl 4-boronobenzoate, p-ethoxycarbonylphenylboronic acid, ethyl 4-dihydroxyboranyl benzoate, 4-carbethoxyphenylboronic acid, benzoic acid, 4-borono-, 1-ethyl ester
Formula Weight 193.99
Physical Form Crystalline Powder
Chemical Name or Material 4-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)