Learn More
4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific™
$1,500.73
Chemical Identifiers
| CAS | 859169-20-3 |
|---|---|
| Molecular Formula | C16H23BO4 |
| Molecular Weight (g/mol) | 290.16 |
| InChI Key | RPRBNRDIESZHFL-UHFFFAOYSA-N |
| Synonym | ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester |
| PubChem CID | 46737998 |
| IUPAC Name | ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC445000050
|
N/A | 5g |
Each for $1,500.73
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 859169-20-3 | |
| 290.16 | |
| ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester | |
| ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
| C16H23BO4 | |
| RPRBNRDIESZHFL-UHFFFAOYSA-N | |
| 46737998 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)OCC |
Specifications
| 859169-20-3 | |
| C16H23BO4 | |
| RPRBNRDIESZHFL-UHFFFAOYSA-N | |
| ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate | |
| 46737998 | |
| 97% |
| 96% min. (GC) | |
| ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)OCC | |
| 290.16 | |
| 290.16 | |
| 4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only