missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58248AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58248AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58248AD Supplier Thermo Scientific Chemicals Supplier No. H58248AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 265330-98-1 | C7H13BrO2Zn | 274.463 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Store at 4°C. Incompatible with oxidizing agents, acid chlorides, acids. Store under dry inert gas. Air Sensitive. Store away from air.

Chemical Identifiers

CAS 265330-98-1
Molecular Formula C7H13BrO2Zn
Molecular Weight (g/mol) 274.463
MDL Number MFCD00671978
InChI Key JJSSUZYKRQXNFR-UHFFFAOYSA-M
Synonym 5-ethoxy-5-oxopentylzinc bromide solution, 5-ethoxy-5-oxopentylzinc bromide, bromozinc 1+ ,ethyl pentanoate, ethyl 5-bromozincio pentanoate, 5-ethoxy-5-oxopentylzinc bromide 0.5 m in tetrahydrofuran, 5-ethoxy-5-oxopentylzinc bromide solution, 0.5 m in thf, 4-ethoxycarbonyl butylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 3260042
IUPAC Name bromozinc(1+);ethyl pentanoate
SMILES CCOC(=O)CCC[CH2-].[Zn+]Br

Specifications

Density 0.978
Linear Formula CH3CH2O2C(CH2)4ZnBr
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 274.46
Chemical Name or Material 4-(Ethoxycarbonyl)butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H261-H314-H351-H335
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only