missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetophenone, 98%

Catalog No. p-7081178
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1103106 5 g
AAA1103114 25 g
AAA1103122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1103106 Supplier Thermo Scientific Chemicals Supplier No. A1103106
Only null left

CAS: 1676-63-7 | C10H12O2 | 164.20 g/mol

4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

Solubility
Soluble in alcohol, water(791.1 mg/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 35°C to 38°C
Boiling Point 145°C to 146°C (13 mmHg)
Flash Point 134°C (273°F)
Quantity 5 g
Beilstein 636783
Solubility Information Soluble in alcohol,water(791.1mg/L).
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4'-Ethoxyacetophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-825-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only