missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxy-3-methoxybenzaldehyde, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 120-25-2 |
|---|---|
| Molecular Formula | C10H12O3 |
| Molecular Weight (g/mol) | 180.2 |
| MDL Number | MFCD00016614 |
| InChI Key | BERFDQAMXIBOHM-UHFFFAOYSA-N |
| Synonym | 4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648 |
| PubChem CID | 67116 |
| IUPAC Name | 4-ethoxy-3-methoxybenzaldehyde |
| SMILES | CCOC1=C(C=C(C=C1)C=O)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC117910250
|
N/A | 25g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C10H12O3 | |
| MFCD00016614 | |
| 4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648 | |
| 4-ethoxy-3-methoxybenzaldehyde |
Specifications
| 120-25-2 | |
| 100.0 | |
| 110°C | |
| 95% | |
| C2H5OC6H3(OCH3)CHO | |
| 08,256 | |
| Solubility in water: slightly soluble | |
| CCOC1=C(C=C(C=C1)C=O)OC | |
| 180.2 | |
| 180.2 | |
| 4-Ethoxy-3-methoxybenzaldehyde, 95% |
| 94.0 | |
| 168.0°C | |
| Authentic | |
| C10H12O3 | |
| MFCD00016614 | |
| 4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648 | |
| BERFDQAMXIBOHM-UHFFFAOYSA-N | |
| 4-ethoxy-3-methoxybenzaldehyde | |
| 67116 | |
| 95% |
Safety and Handling
EINECSNumber : 204-382-1
TSCA : TSCA
RUO – Research Use Only