missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Epichlortetracycline hydrochloride, can be used as secondary standard

4-Epichlortetracycline hydrochloride, CAS # 14297-93-9, is a broad-spectrum, bacteriostatic antibiotic of the tetracycline antibiotic family.

$306.55 - $1120.12

Chemical Identifiers

CAS 101342-45-4
Molecular Formula C22H23ClN2O8·ClH
Molecular Weight (g/mol) 515.34
MDL Number MFCD09842537
InChI Key QYAPHLRPFNSDNH-PNEIEYGESA-N
Synonym epichlortetracycline hydrochloride
PubChem CID 92043146
IUPAC Name (4R,4aS,5aS,6R,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC268231000
SDS View Documents
Thermo Scientific Chemicals
268231000
100 mg Glass bottle
Each for $306.55
Only null left
 
AC268235000
SDS View Documents
Thermo Scientific Chemicals
268235000
500 mg Glass bottle
Each for $1,120.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• 4-Epichlortetracycline is the hydrochloride salt of 4-Epichlortetracycline, a tetracycline ring structure with a chloride substitution

• 4-Epichlortetracycline hydrochloride (4-Epi-chlortetracycline) is an epimer of chlortetracycline

• 4-Epichlortetracycline suppresses neutrophil activity and dampens the immune response

• Acts as an aminoacyl-tRNA analog to disrupt polypeptide synthesis, reducing bacterial growth Applications

• Can be used for in vitro research on cancer and inflammatory mechanisms

• Intended for use as a selective agent and secondary standard

Specifications

Chemical Identifiers

101342-45-4
515.34
QYAPHLRPFNSDNH-PNEIEYGESA-N
92043146
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
C22H23ClN2O8·ClH
MFCD09842537
epichlortetracycline hydrochloride
(4R,4aS,5aS,6R,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride

Specifications

101342-45-4
100.0
Orange to Yellow
80% min. (as is,calculated)
C22H23ClN2O8·ClH
100 mg
Solubility in water: 67g/L. Other solubilities: 20g/L methanol
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
515.34
515.34
Powder
96.0
250.0°C
Authentic
Glass bottle
MFCD09842537
epichlortetracycline hydrochloride
QYAPHLRPFNSDNH-PNEIEYGESA-N
(4R,4aS,5aS,6R,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
92043146
≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
4-Epichlortetracycline hydrochloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Suspected of damaging the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only