missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecyldiethylenetriamine, 80%, tech.

Catalog No. AC388210250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC388210250 Supplier Thermo Scientific Chemicals Supplier No. 388210250

CAS: 4182-44-9 | C16H37N3 | 271.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4182-44-9
Molecular Formula C16H37N3
Molecular Weight (g/mol) 271.48
MDL Number MFCD00041897
InChI Key BRNPMBFYMPUNSK-UHFFFAOYSA-N
Synonym 4-dodecyldiethylenetriamine, unii-vg94uin7ys, 1,2-ethanediamine, n-2-aminoethyl-n-dodecyl, n-2-aminoethyl-n-dodecyl-1,2-ethanediamine, vg94uin7ys, n1-2-aminoethyl-n1-dodecyl-1,2-ethanediamine, 1,2-ethanediamine, n1-2-aminoethyl-n1-dodecyl, n-2-aminoethyl-n-dodecylethylenediamine, bis 2-aminoethyl dodecyl amine, n'-2-aminoethyl-n'-dodecylethane-1,2-diamine
PubChem CID 77847
IUPAC Name N'-(2-aminoethyl)-N'-dodecylethane-1,2-diamine
SMILES CCCCCCCCCCCCN(CCN)CCN

Specifications

CAS Min % 75.0
CAS Max % 100.0
Melting Point 22°C
Color Colorless to Yellow
Boiling Point 140°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Refractive Index 1.4640 to 1.468
Linear Formula CH3(CH2)11N(CH2CH2NH2)2
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,chloroform,benzene,toluene,,diethyl ether
Formula Weight 271.48
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material 4-Dodecyldiethylenetriamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-056-2

RUO – Research Use Only