missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™

Catalog No. D42911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42911G 1 g
D42915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42911G Supplier TCI America Supplier No. D42911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1173788-58-3
Molecular Formula C22H39BO2S
Molecular Weight (g/mol) 378.42
MDL Number MFCD22124727
InChI Key KFTOUTPAXZWQLP-UHFFFAOYSA-N
Synonym 2-(4-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dodecyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 71306582
IUPAC Name 2-(4-dodecylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCCCCCCCCCCCC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 378.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene