missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D35371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35371G Supplier TCI America Supplier No. D35371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(Diphenylamino)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 201802-67-7
Melting Point 228°C
Color Green-White
Molecular Formula C18H16BNO2
Quantity 1 g
MDL Number MFCD06798117
Synonym 4-diphenylamino phenylboronic acid, triphenylamine-4-boronic acid, 4-diphenylamino phenyl boronic acid, 4-diphenylamino benzeneboronic acid, 4-n,n-diphenylamino phenylboronic acid, boronic acid, b-4-diphenylamino phenyl, 4-n-diphenylamino phenylboronic acid, 4-diphenylamino phenylboronicacid, boronic acid, 4-diphenylamino phenyl
InChI Key TWWQCBRELPOMER-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3)(O)O
IUPAC Name [4-(N-phenylanilino)phenyl]boronic acid
Molecular Weight (g/mol) 289.141
PubChem CID 12166934
Formula Weight 289.14
Physical Form Crystalline Powder
Show More Show Less