missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridinium Bromide Perbromide 97.0+%, TCI America™

Catalog No. D17875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17875G 5 g
D178725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17875G Supplier TCI America Supplier No. D17875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92976-81-3
Molecular Formula C7H10Br3N2-
Molecular Weight (g/mol) 361.883
MDL Number MFCD00043439
InChI Key TYDZHQMKSLAFKM-UHFFFAOYSA-N
Synonym 4-dimethylamino pyridine hydrobromide perbromide
PubChem CID 131863682
SMILES CN(C)C1=CC=NC=C1.Br[Br-]Br

Specifications

Melting Point 131°C
Quantity 5 g
UN Number 3261
Formula Weight 362.89
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminopyridinium Bromide Perbromide