missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylaminocinnamaldehyde 98.0+%, TCI America™
Supplier: TCI America D06485G
Specifications
| 4-Dimethylaminocinnamaldehyde | |
| 140°C | |
| MFCD00007002 | |
| 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent | |
| CN(C)C1=CC=C(C=C1)C=CC=O | |
| 175.231 | |
| 175.23 | |
| Crystalline Powder |
| 6203-18-5 | |
| C11H13NO | |
| 5 g | |
| RUKJCCIJLIMGEP-ONEGZZNKSA-N | |
| (E)-3-[4-(dimethylamino)phenyl]prop-2-enal | |
| 5284506 | |
| ≥98.0% (T) |
Chemical Identifiers
| 6203-18-5 | |
| 175.231 | |
| RUKJCCIJLIMGEP-ONEGZZNKSA-N | |
| 5284506 | |
| CN(C)C1=CC=C(C=C1)C=CC=O |
| C11H13NO | |
| MFCD00007002 | |
| 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent | |
| (E)-3-[4-(dimethylamino)phenyl]prop-2-enal |
Safety and Handling
EINECSNumber : (3)-1187
TSCA : Yes