missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminocinnamaldehyde, 98%

Catalog No. AAB2474114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2474114 25 g
AAB2474106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2474114 Supplier Thermo Scientific Chemicals Supplier No. B2474114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6203-18-5 | C11H13NO | 175.231 g/mol

4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

Solubility
Soluble in chloroform, ethanol, dioxane.

Notes
Air & light sensitive. Store at -20°C. Store in dark. Incompatible with oxidizing agents, air, light.

Chemical Identifiers

CAS 6203-18-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00007002
InChI Key RUKJCCIJLIMGEP-ONEGZZNKSA-N
Synonym 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent
PubChem CID 5284506
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enal
SMILES CN(C)C1=CC=C(C=C1)C=CC=O

Specifications

Melting Point 137°C to 144°C
Quantity 25 g
Beilstein 972369
Sensitivity Air and light sensitive
Solubility Information Soluble in chloroform,ethanol,dioxane.
Formula Weight 175.23
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminocinnamaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-267-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only